Isobutyryl chloride ≥98%
Supplier: BeanTown Chemical
135185-100G
135185-500G
BT135185-100GEA
38.25
USD
BT135185-100G
BT135185-500G
Isobutyryl chloride ≥98%
Isobutyryl chloride
CAS: 79-30-1; EC No: 201-194-1; MDL No: MFCD00000717; RTECS: UC3944000
UN No: UN2395; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: (CH3)2CHCOCl; Molecular Formula: C4H7ClO; MW: 106.55
Melting Point: -90°; Boiling Point: 91-93°; Flash point: 8°C (46°F)
Density (g/mL): 1.017; Refractive Index: 1.407
Moisture Sensitive
- RTECS: UC3944000
- Refractive index: 1.407
- Moisture Sensitive
- Liquid
Formula:
C₄H₇ClO MW: 106.55 g/mol Boiling Pt: 91…93 °C Melting Pt: –90 °C Density: 1.017 g/mL Flash Pt: 8 °C (46 °F) |
MDL Number:
MFCD00000717 CAS Number: 79-30-1 EINECS: 201-194-1 UN: 2395 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...