Lithium fluoride ≥98%

Supplier: BeanTown Chemical

135155-100G 135155-2.5KG 135155-500G
BT135155-100GEA 135.72 USD
BT135155-100G BT135155-2.5KG BT135155-500G
Lithium fluoride ≥98%
Lithium fluoride

CAS: 7789-24-4; EC No: 232-152-0; MDL No: MFCD00011090; RTECS: OJ6125000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: LiF; MW: 25.94 Melting Point: 845°; Boiling Point: 1673° Density (g/mL): 2.64


  • RTECS: OJ6125000
  • Solid

Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1673 °C
Melting Pt: 845 °C
Density: 2.64 g/mL
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR