Barium chloride dihydrate ACS

Supplier: BeanTown Chemical

Synonyms: Barium dichloride

135125-2KG 135125-500G
BT135125-2KGEA 139.86 USD
BT135125-2KG BT135125-500G
Barium chloride dihydrate ACS
Barium chloride dihydrate

CAS: 10326-27-9; EC No: 233-788-1; MDL No: MFCD00149154; RTECS: CQ8751000 UN No: UN1564; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: BaCl2·2H2O; MW: 244.26


  • RTECS: CQ8751000
  • Crystalline

Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR