Nickel(II) bromide trihydrate ≥98%

Supplier: BeanTown Chemical

135115-250G 135115-50G
BT135115-250GEA 405 USD
BT135115-250G BT135115-50G
Nickel(II) bromide trihydrate ≥98%
Nickel(II) bromide trihydrate

CAS: 7789-49-3; EC No: 236-665-0; MDL No: MFCD00011141 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Granular; Linear Formula: NiBr2·3H2O; MW: 272.58 Hygroscopic


  • Hygroscopic
  • Crystalline/Granular

Formula: Br₂Ni·3H₂O
MDL Number: MFCD00011141
CAS Number: 7789-49-3
EINECS: 236-665-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR