4-Anisoyl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Methoxybenzoyl chloride

135105-100G 135105-25G
BT135105-100GEA 114.98 USD
BT135105-100G BT135105-25G
4-Anisoyl chloride ≥97%
4-Anisoyl chloride

CAS: 100-07-2; EC No: 202-816-4; MDL No: MFCD00000687 UN No: UN1729; Haz Class: 8; Packing Group: II Liquid/Low Melting Solid; Molecular Formula: C8H7ClO2; MW: 170.59 Melting Point: 22°; Boiling Point: 262-263°; Flash point: 87°C (189°F) Density (g/mL): 1.26; Refractive Index: 1.581 Moisture Sensitive


  • Refractive index: 1.581
  • Moisture Sensitive
  • Liquid/Low Melting Solid

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 263
Melting Pt: 22 °C
Density: 1.26 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00000687
CAS Number: 100-07-2
EINECS: 202-816-4
UN: 1729
ADR: 8 II
Merck Index: 13,00676

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR