Barium peroxide, anhydrous 84

Supplier: BeanTown Chemical

135090-1KG 135090-250G 135090-50G
BT135090-1KGEA 94 USD
BT135090-1KG BT135090-250G BT135090-50G
Barium peroxide, anhydrous 84
Barium peroxide

CAS: 1304-29-6; EC No: 215-128-4; MDL No: MFCD00003454 UN No: UN1449; Haz Class: 5.1 (6.1); Packing Group: II Powder; Molecular Formula: BaO2; MW: 169.34


  • Powder

Formula: BaO₂
MW: 169.33 g/mol
Melting Pt: 450
MDL Number: MFCD00003454
CAS Number: 1304-29-6
EINECS: 215-128-4
UN: 1449
ADR: 5.1 II
Merck Index: 12,01018

Order Now

Specification Test Results

Assay 84% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR