2-(2-Methoxyphenoxy)ethylamine ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-(2-Aminoethoxy)-2-methoxybenzene
135075-1G
135075-250MG
BT135075-1GEA
309
USD
BT135075-1G
BT135075-250MG
2-(2-Methoxyphenoxy)ethylamine ≥97%
2-(2-Methoxyphenoxy)ethylamine
CAS: 1836-62-0; MDL No: MFCD00235185
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C9H13NO2 ; MW: 167.21
Density (g/mL): 1.11
Air Sensitive
- Air Sensitive
- Liquid
Formula:
C₉H₁₃NO₂ Density: 1.11 g/mL |
MDL Number:
MFCD00235185 CAS Number: 1836-62-0 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...