Vanadium(V) oxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Vanadium pentoxide, di-Vanadium pentaoxide
135045-1KG
135045-250G
BT135045-1KGEA
159
USD
BT135045-1KG
BT135045-250G
Vanadium(V) oxide ≥99%
Vanadium(V) oxide
CAS: 1314-62-1; EC No: 215-239-8; MDL No: MFCD00011457; RTECS: YW2125000
UN No: UN2862; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: V2O5; MW: 181.88
Melting Point: 690°; Boiling Point: 1750° (decomposes)
Density (g/mL): 3.357
- RTECS: YW2125000
- Powder
Formula:
O₅V₂ MW: 181.88 g/mol Boiling Pt: 1750 °C (dec.) Melting Pt: 690 °C Density: 3.357 g/mL |
MDL Number:
MFCD00011457 CAS Number: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...