Copper(I) oxide ≥99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Cuprous oxide, Dicopper oxide
135000-100G
135000-25G
135000-5G
BT135000-100GEA
239
USD
BT135000-100G
BT135000-25G
BT135000-5G
Copper(I) oxide ≥99% (trace metals basis)
Copper(I) oxide
CAS: 1317-39-1; EC No: 215-270-7; MDL No: MFCD00010974; RTECS: GL8050000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: Cu2O; MW: 143.08
Melting Point: 1232°
Density (g/mL): 6
Air Sensitive, Moisture Sensitive
- RTECS: GL8050000
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
Cu₂O MW: 143.09 g/mol Melting Pt: 1232 °C Density: 6 g/mL |
MDL Number:
MFCD00010974 CAS Number: 1317-39-1 EINECS: 215-270-7 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...