2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98%

Supplier: BeanTown Chemical

Synonyms: TEMPO, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethylpiperidine 1-oxyl

134995-1G 134995-25G 134995-5G
BT134995-1GEA 27.73 USD
BT134995-1G BT134995-25G BT134995-5G
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98%
2,2,6,6-Tetramethylpiperidinooxy (TEMPO)

CAS: 2564-83-2; EC No: 219-888-8; MDL No: MFCD00009599; RTECS: TN8991900 UN No: UN3263; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C9H18NO; MW: 156.25 Melting Point: 36-38°; Boiling Point: 193° (decomposes); Flash point: 67°C (152°F)


  • RTECS: TN8991900
  • Solid

Formula: C₉H₁₈NO
MW: 156.25 g/mol
Boiling Pt: 193 °C (dec.)
Melting Pt: 36…38 °C
Flash Pt: 67 °C (152 °F)
MDL Number: MFCD00009599
CAS Number: 2564-83-2
EINECS: 219-888-8
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR