Lithium hydride ≥99% (trace metals basis)
Supplier: BeanTown Chemical
134985-100G
134985-25G
BT134985-100GEA
249.5
USD
BT134985-100G
BT134985-25G
Lithium hydride ≥99% (trace metals basis)
Lithium hydride
CAS: 7580-67-8; EC No: 231-484-3; MDL No: MFCD00011074; RTECS: OJ6300000
UN No: UN1414; Haz Class: 4.3; Packing Group: I
Powder; Linear Formula: LiH; MW: 7.95
Melting Point: 680°
Density (g/mL): 0.82
Moisture Sensitive
- RTECS: OJ6300000
- Moisture Sensitive
- Powder
Formula:
HLi MW: 7.95 g/mol Melting Pt: 680 °C Density: 0.82 g/mL |
MDL Number:
MFCD00011074 CAS Number: 7580-67-8 EINECS: 231-484-3 UN: 1414 ADR: 4.3,I |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...