2,3,4,5,6-Pentachloroaniline ≥96%
Supplier: BeanTown Chemical
CAS: 527-20-8; EC No: 208-410-3; MDL No: MFCD00014769; RTECS: BY7910000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Linear Formula: C6Cl5NH2; MW: 265.35
Melting Point: 232-234°; Flash point:
- RTECS: BY7910000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₆H₂Cl₅N Melting Pt: 232…234 °C Flash Pt: >100 °C (>212 °F) |
MDL Number:
MFCD00014769 CAS Number: 527-20-8 EINECS: 208-410-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...