1,2-Bis(2-chloroethoxy)ethane ≥97%
Supplier: BeanTown Chemical
134920-100G
134920-500G
BT134920-100GEA
54.66
USD
BT134920-100G
BT134920-500G
1,2-Bis(2-chloroethoxy)ethane ≥97%
1,2-Bis(2-chloroethoxy)ethane
CAS: 112-26-5; EC No: 203-952-7; MDL No: MFCD00000976; RTECS: KH4900000
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: ClCH2CH2OCH2CH2OCH2CH2Cl ; Molecular Formula: C6H12Cl2O2; MW: 187.06
Boiling Point: 235°; Flash point: 121°C (250°F)
Density (g/mL): 1.197; Refractive Index: 1.461
- RTECS: KH4900000
- Refractive index: 1.461
- Liquid
Formula:
ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl MW: 187.07 g/mol Boiling Pt: 235 °C Density: 1.197 g/mL Flash Pt: 121 °C (250 °F) |
MDL Number:
MFCD00000976 CAS Number: 112-26-5 EINECS: 203-952-7 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...