1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Methylbis(trimethylsilyloxy)silane
134910-250ML
134910-50ML
BT134910-250MLEA
403
USD
BT134910-250ML
BT134910-50ML
1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥97%
1,1,1,3,5,5,5-Heptamethyltrisiloxane
CAS: 1873-88-7; EC No: 217-496-1; MDL No: MFCD00048000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C7H22O2Si3; MW: 222.50
Boiling Point: 142°; Flash point: 22°C (72°F)
Density (g/mL): 0.846; Refractive Index: 1.459
- Refractive index: 1.459
- Liquid
Formula:
C₇H₂₂O₂Si₃ MW: 222.51 Boiling Pt: 142 °C Density: 0.846 g/mL Flash Pt: 22 °C (72 °F) |
MDL Number:
MFCD00048000 CAS Number: 1873-88-7 EINECS: 217-496-1 ADR: 3 II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...