Formic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: Aminic Acid, Methanoic acid

134900-10KG 134900-2.5KG 134900-500G
BT134900-10KGEA 306 USD
BT134900-10KG BT134900-2.5KG BT134900-500G
Formic acid ≥97%
Formic acid

CAS: 64-18-6; EC No: 200-579-1; MDL No: MFCD00003297; RTECS: LQ4900000 UN No: UN1779; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: HCOOH; MW: 46.03 Melting Point: 8°; Boiling Point: 100-101°; Flash point: 48°C (119°F) Density (g/mL): 1.22; Refractive Index: 1.371


  • RTECS: LQ4900000
  • Refractive index: 1.371
  • Liquid

Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100…101 °C
Melting Pt: 8 °C
Density: 1.22 g/mL
Flash Pt: 48 °C (119 °F)
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR