Tributylphosphine ≥95%

Supplier: BeanTown Chemical

Synonyms: Tri-n-butylphosphine, TBP

134895-100ML 134895-25ML 134895-500ML
BT134895-100MLEA 53.3 USD
BT134895-100ML BT134895-25ML BT134895-500ML
Tributylphosphine ≥95%
Tributylphosphine

CAS: 998-40-3; EC No: 213-651-2; MDL No: MFCD00009462; RTECS: SZ3270000 UN No: UN3254; Haz Class: 4.2; Packing Group: I Liquid; Linear Formula: [CH3(CH2)3]3P; Molecular Formula: C12H27P; MW: 202.32 Melting Point: -60°; Boiling Point: 150°/50 mmHg; Flash point: 37°C (99°F) Density (g/mL): 0.810; Refractive Index: 1.462 Air Sensitive, Moisture Sensitive


  • RTECS: SZ3270000
  • Refractive index: 1.462
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 240
Melting Pt: -65
Density: 0.810 g/mL
Flash Pt: 37 °C (99 °F)
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 3254
ADR: 4.2 I

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR