Potassium bromate ≥99%
Supplier: BeanTown Chemical
134870-100G
134870-2.5KG
134870-500G
BT134870-100GEA
45.59
USD
BT134870-100G
BT134870-2.5KG
BT134870-500G
Potassium bromate ≥99%
Potassium bromate
CAS: 7758-01-2; EC No: 231-829-8; MDL No: MFCD00011359; RTECS: EF8725000
UN No: UN1484; Haz Class: 5.1; Packing Group: II
Granular; Linear Formula: KBrO3; MW: 167.00
Melting Point: 350°
Density (g/mL): 3.27
- RTECS: EF8725000
- Granular
Formula:
KBrO₃ Melting Pt: 350 °C Density: 3.27 g/mL |
MDL Number:
MFCD00011359 CAS Number: 7758-01-2 EINECS: 231-829-8 UN: 1484 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...