Pivaloylacetonitrile ≥99%

Supplier: BeanTown Chemical

Synonyms: 4,4-Dimethyl-3-oxopentanenitrile, 4,4-Dimethyl-3-oxovaleronitrile, Trimethylacetylacetonitrile

134845-100G 134845-25G 134845-500G
BT134845-100GEA 199 USD
BT134845-100G BT134845-25G BT134845-500G
Pivaloylacetonitrile ≥99%
Pivaloylacetonitrile

CAS: 59997-51-2; EC No: 262-017-1; MDL No: MFCD00010208; RTECS: SA2991000 UN No: UN3439; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: (CH3)3CCOCH2CN; Molecular Formula: C7H11NO; MW: 125.17 Melting Point: 66-69°; Boiling Point: 125-126°/22 mmHg


  • RTECS: SA2991000
  • Crystalline

Formula: C₇H₁₁NO
MW: 125.17 g/mol
Boiling Pt: 125…126 °C/22 mmHg
Melting Pt: 66…69 °C
MDL Number: MFCD00010208
CAS Number: 59997-51-2
EINECS: 262-017-1
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR