Magnesium nitrate hexahydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
134835-2KG
134835-500G
134835-50G
BT134835-2KGEA
144.29
USD
BT134835-2KG
BT134835-500G
BT134835-50G
Magnesium nitrate hexahydrate 98.0-102.0% ACS
Magnesium nitrate hexahydrate
CAS: 13446-18-9; EC No: 233-826-7; MDL No: MFCD00149779; RTECS: OM3756000
UN No: UN1474; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Mg(NO3)2·6H2O ; MW: 256.41
Melting Point: 89° (decomposes); Boiling Point: 330°
Density (g/mL): 1.636
Hygroscopic
- RTECS: OM3756000
- Hygroscopic
- Crystalline
Formula:
Mg(NO₃)₂·6H₂O Boiling Pt: 330 °C Melting Pt: 89 °C (dec.) Density: 1.636 g/mL |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III |
Specification Test Results
Assay | 98.0-102.0% |
pH of 5% solution at 25°C | 5.0-8.2 |
Insoluble matter | ≤0.005% |
Chloride (Cl) | ≤0.001% |
Phosphate (PO4) | ≤5ppm |
Sulfate (SO4) | ≤0.005% |
Ammonium (NH4) | ≤0.003% |
Barium (Ba) | ≤0.005% |
Calcium (Ca) | ≤0.01% |
Manganese (Mn) | ≤5ppm |
Potassium (K) | ≤0.005% |
Sodium (Na) | ≤0.005% |
Strontium (Sr) | ≤0.005% |
Heavy metals | ≤5ppm |
Iron (Fe) | ≤5ppm |
Copper (Cu) | ≤0.005% |
Nitrate (NO3) | ≤0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...