Terbium(III) nitrate hydrate ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Terbium(3+) nitrate hydrate (1:3:1)
134820-25G
134820-5G
BT134820-25GEA
208.78
USD
BT134820-25G
BT134820-5G
Terbium(III) nitrate hydrate ≥99.9% (trace rare earth metals basis)
Terbium(III) nitrate hydrate
EC No: 233-138-7; MDL No: MFCD00011257
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Linear Formula: Tb(NO3)3·xH2O; MW: 344.93 (anhydrous)
- Crystalline Aggregates
Formula:
Tb(NO₃)₃·xH₂O MW: 344.94 g/mol (anhy) |
MDL Number:
MFCD00011257 CAS Number: 94219-56-4 EINECS: 233-138-7 ADR: 5.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...