3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥98%

Supplier: BeanTown Chemical

Synonyms: Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol

134815-1G 134815-25G 134815-5G
BT134815-1GEA 20 USD
BT134815-1G BT134815-25G BT134815-5G
3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥98%
3,4-Dihydroxy-3-cyclobutene-1,2-dione

CAS: 2892-51-5; EC No: 220-761-4; MDL No: MFCD00001334; RTECS: GU1800000 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C4H2O4; MW: 114.06 Melting Point: 300°


  • RTECS: GU1800000
  • Crystalline

Formula: C₄H₂O₄
Melting Pt: >300 °C
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR