Hexachloroplatinic(IV) acid hydrate ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate

134810-1G 134810-5G
BT134810-1GEA 170.08 USD
BT134810-1G BT134810-5G
Hexachloroplatinic(IV) acid hydrate ≥99.999% (trace metals basis)
Hexachloroplatinic(IV) acid hydrate

CAS: 26023-84-7; EC No: 241-010-7; MDL No: MFCD00149909; RTECS: TP1500000 UN No: UN2507; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: H2PtCl6·xH2O; MW: 409.82 (anhydrous) Melting Point: 60° (decomposes) Hygroscopic


  • RTECS: TP1500000
  • Hygroscopic
  • Crystalline

Formula: H₂Cl₆Pt·H₂O
MW: 427.83 g/mol
Melting Pt: 60 °C (dec.)
MDL Number: MFCD00149909
CAS Number: 26023-84-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR