Gadolinium(III) isopropoxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Gadolinium(III) i-propoxide
134800-1G
134800-5G
BT134800-1GEA
176
USD
BT134800-1G
BT134800-5G
Gadolinium(III) isopropoxide ≥99%
Gadolinium(III) isopropoxide
CAS: 14532-05-9; MDL No: MFCD00144381
UN No: UN2925; Haz Class: 4.1 (8); Packing Group: III
Powder; Molecular Formula: C9H21GdO3 ; MW: 334.51
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
C₉H₂₁GdO₃ MW: 334.514 g/mol |
MDL Number:
MFCD00144381 CAS Number: 14532-05-9 ADR: 4.1 (8) III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...