Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (trace metals basis)
Supplier: BeanTown Chemical
134785-25ML
134785-5ML
BT134785-25MLEA
176
USD
BT134785-25ML
BT134785-5ML
Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (trace metals basis)
Tin(IV) isopropoxide
CAS: 1184-61-8; MDL No: MFCD00145408
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: Sn[OCH(CH3)2]4; Molecular Formula: C12H28O4Sn; MW: 355.06
Boiling Point: 83°
Density (g/mL): 0.839; Refractive Index: 1.394
Moisture Sensitive
- Refractive index: 1.394
- Moisture Sensitive
- Liquid
Formula:
C₁₂H₂₈O₄Sn MW: 355.062 g/mol Boiling Pt: 83 °C Density: 0.839 g/mL |
MDL Number:
MFCD00145408 CAS Number: 1184-61-8 ADR: 3 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...