Cyclopropylméthanol ≥98%
Supplier: BeanTown Chemical
Synonyms:
Cyclopropyl carbinol, Cyclopropanemethanol
134780-100G
134780-25G
134780-5G
BT134780-100GEA
159
USD
BT134780-100G
BT134780-25G
BT134780-5G
Cyclopropylméthanol ≥98%
Cyclopropylméthanol
CAS: 2516-33-8; EC No: 219-735-5; MDL No: MFCD00001309; RTECS: GZ2212250
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C4H8O; MW: 72.11
Boiling Point: 123-124°/738 mmHg; Flash point: 35°C (95°F)
Density (g/mL): 0.89; Refractive Index: 1.431
- RTECS: GZ2212250
- Refractive index: 1.431
- Liquid
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 123…124 °C/738 mmHg Density: 0.89 g/mL Flash Pt: 35 °C (95 °F) |
MDL Number:
MFCD00001309 CAS Number: 2516-33-8 EINECS: 219-735-5 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...