1-Phenyl-2-thiourea ≥97%

Supplier: BeanTown Chemical

Synonyms: N-Phenylthiourea, 1-Phenylthiourea

134715-10G 134715-250G 134715-50G
BT134715-10GEA 40.02 USD
BT134715-10G BT134715-250G BT134715-50G
1-Phenyl-2-thiourea ≥97%
1-Phenyl-2-thiourea

CAS: 103-85-5; EC No: 203-151-2; MDL No: MFCD00004933; RTECS: YU1400000 UN No: UN2811; Haz Class: 6.1; Packing Group: I Solid; Linear Formula: C6H5NHCSNH2; Molecular Formula: C7H8N2S; MW: 152.21 Melting Point: 145-150°


  • RTECS: YU1400000
  • Solid

Formula: C₇H₈N₂S
MW: 152.22 g/mol
Melting Pt: 145…150 °C
MDL Number: MFCD00004933
CAS Number: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR