2-Chloroethyl ethyl ether ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Ethoxyethyl chloride, 2-Chlorodiethyl ether, 2-Chloroethylethylether
134700-100G
134700-25G
BT134700-100GEA
253
USD
BT134700-100G
BT134700-25G
2-Chloroethyl ethyl ether ≥98%
2-Chloroethyl ethyl ether
CAS: 628-34-2; EC No: 211-037-9; MDL No: MFCD00000974; RTECS: KN6250000
UN No: UN3271; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2OCH2CH2Cl; Molecular Formula: C4H9ClO; MW: 108.57
Boiling Point: 107°; Flash point: 6°C (43°F)
Density (g/mL): 0.989; Refractive Index: 1.411
- RTECS: KN6250000
- Refractive index: 1.411
- Liquid
Formula:
C₄H₉ClO MW: 108.57 g/mol Boiling Pt: 107 °C Density: 0.989 g/mL Flash Pt: 6 °C (43 °F) |
MDL Number:
MFCD00000974 CAS Number: 628-34-2 EINECS: 211-037-9 UN: 3271 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...