Praseodymium(III) nitrate hexahydrate ≥99.99% (trace rare earth metals basis)

Supplier: BeanTown Chemical

Synonyms: Praseodymium trinitrate hexahydrate

134650-100G 134650-25G
BT134650-100GEA 556 USD
BT134650-100G BT134650-25G
Praseodymium(III) nitrate hexahydrate ≥99.99% (trace rare earth metals basis)
Praseodymium(III) nitrate hexahydrate

CAS: 15878-77-0; EC No: 233-796-5; MDL No: MFCD00149826 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Pr(NO3)3·6H2O; MW: 435.01 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: N₃O₉Pr·6H₂O
MW: 435.01 g/mol
MDL Number: MFCD00149826
CAS Number: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR