3-Amino-4-chloropyridine ≥98%
Supplier: BeanTown Chemical
134630-1G
134630-5G
BT134630-1GEA
89
USD
BT134630-1G
BT134630-5G
3-Amino-4-chloropyridine ≥98%
3-Amino-4-chloropyridine
CAS: 20511-15-3; MDL No: MFCD00235124
UN No: UN2811 ; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C5H5ClN2; MW: 128.56
Melting Point: 58-63°
- Crystalline
Formula:
C₅H₅ClN₂ Melting Pt: 58…63 °C |
MDL Number:
MFCD00235124 CAS Number: 20511-15-3 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...