Trityl chloride resin 0,5 - 2,5 mmol/g on poly(styrene-divinylbenzene), 1% cross-linked
Supplier: BeanTown Chemical
Synonyms:
Trityl chloride, polymer-supported, Chlorodiphenylmethyl Polystyrene resin
134605-1G
134605-25G
134605-5G
BT134605-1GEA
39
USD
BT134605-1G
BT134605-25G
BT134605-5G
Trityl chloride resin 0,5 - 2,5 mmol/g on poly(styrene-divinylbenzene), 1% cross-linked
Trityl chloride resin
MDL No: MFCD00284810
UN No: UN3261; Haz Class: 8; Packing Group: III
100-200 Mesh Beads
Moisture Sensitive
- Moisture Sensitive
- 100-200 Mesh Beads
MDL Number:
MFCD00284810 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...