meta-Phosphoric acid 33.5-36.5% (w/w) balance NaPO3 stabilizer ACS

Supplier: BeanTown Chemical

Synonyms: Metaphosphoric acid

134565-100G 134565-25G 134565-500G
BT134565-100GEA 106.13 USD
BT134565-100G BT134565-25G BT134565-500G
meta-Phosphoric acid 33.5-36.5% (w/w) balance NaPO3 stabilizer ACS
meta-Phosphoric acid

CAS: 37267-86-0; EC No: 253-433-4; MDL No: MFCD00011351 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: HPO3; MW: 79.98 Melting Point: 200°; Boiling Point: 600°/1,013 mmHg Density (g/mL): 2 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: (HPO₃)n
MW: 79.98 g/mol
Boiling Pt: 600 °C/1,013 mmHg
Melting Pt: 200 °C
Density: 2 g/mL
MDL Number: MFCD00011351
CAS Number: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR