Antimony (III) sulfide ≥98%

Supplier: BeanTown Chemical

Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black

134450-1KG 134450-250G
BT134450-1KGEA 133 USD
BT134450-1KG BT134450-250G
Antimony (III) sulfide ≥98%
Antimony (III) sulphide

CAS: 1345-04-6; EC No: 215-713-4; MDL No: MFCD00011217 UN No: UN1549; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: Sb2S3; MW: 339.72 Melting Point: 550°


  • Powder

Formula: S₃Sb₂
MW: 339.72 g/mol
Melting Pt: 550 °C
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR