Triethyl borate ≥97%
Supplier: BeanTown Chemical
134400-100ML
134400-2.5L
134400-500ML
BT134400-100MLEA
59.6
USD
BT134400-100ML
BT134400-2.5L
BT134400-500ML
Triethyl borate ≥97%
Triethyl borate
CAS: 150-46-9; EC No: 205-760-9; MDL No: MFCD00009073; RTECS: ED5075000
UN No: UN1176; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3CH2O)3B; Molecular Formula: C6H15BO3; MW: 145.99
Melting Point: -85°; Boiling Point: 117-118°; Flash point: 11°C (52°F)
Density (g/mL): 0.858; Refractive Index: 1.374
Moisture Sensitive
- RTECS: ED5075000
- Refractive index: 1.374
- Moisture Sensitive
- Liquid
Formula:
C₆H₁₅BO₃ MW: 145.99 g/mol Boiling Pt: 117…118 °C Melting Pt: –85 °C Density: 0.858 g/mL Flash Pt: 11 °C (52 °F) |
MDL Number:
MFCD00012846 CAS Number: 150-46-9 EINECS: 205-760-9 UN: 1176 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...