DL-Camphor ≥96%

Supplier: BeanTown Chemical

Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone

134390-100G 134390-2.5KG 134390-500G
BT134390-100GEA 30.44 USD
BT134390-100G BT134390-2.5KG BT134390-500G
DL-Camphor ≥96%
DL-Camphor

CAS: 76-22-2; EC No: 200-945-0; MDL No: MFCD00074738; RTECS: EX1225000 UN No: UN2717; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: C10H16O; MW: 152.23 Melting Point: 175-177°; Boiling Point: 204°; Flash point: 64°C (147°F) Density (g/mL): 0.992


  • RTECS: EX1225000
  • Solid

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C
Melting Pt: 175…177 °C
Density: 0.992 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR