Ethylbenzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
Phenylethane
134380-100ML
134380-2.5L
134380-500ML
BT134380-100MLEA
32.74
USD
BT134380-100ML
BT134380-2.5L
BT134380-500ML
Ethylbenzene ≥99%
Ethylbenzene
CAS: 100-41-4; EC No: 202-849-4; MDL No: MFCD00011647; RTECS: DA0700000
UN No: UN1175; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H5CH2CH3; Molecular Formula: C8H10; MW: 106.17
Melting Point: -95°; Boiling Point: 136°; Flash point: 15°C (59°F)
Density (g/mL): 0.867; Refractive Index: 1.495
- RTECS: DA0700000
- Refractive index: 1.495
- Liquid
Formula:
C₈H₁₀ MW: 106.17 g/mol Boiling Pt: 136 °C Melting Pt: –95 °C Density: 0.867 g/mL Flash Pt: 15 °C (59 °F) |
MDL Number:
MFCD00011647 CAS Number: 100-41-4 EINECS: 202-849-4 UN: 1175 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...