Aluminium bromide ≥99.997%
Supplier: BeanTown Chemical
Synonyms:
Aluminum bromide
134360-25G
134360-5G
BT134360-25GEA
159
USD
BT134360-25G
BT134360-5G
Aluminium bromide ≥99.997%
Aluminium bromide
CAS: 7727-15-3; EC No: 231-779-7; MDL No: MFCD00003421; RTECS: BD0350000
UN No: UN1725; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: AlBr3; MW: 266.69
Melting Point: 94-98°
Density (g/mL): 3.205
Moisture Sensitive
- RTECS: BD0350000
- Moisture Sensitive
- Crystalline
Formula:
AlBr₃ MW: 266.69 g/mol Boiling Pt: 265 Melting Pt: 97 Density: 3.205 g/mL |
MDL Number:
MFCD00003421 CAS Number: 7727-15-3 EINECS: 231-779-7 UN: 1725 ADR: 8 II Merck Index: 13,00330 |
Specification Test Results
Assay | 99.997% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...