Gallium metal ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
CAS: 7440-55-3; EC No: 231-163-8; MDL No: MFCD00134045; RTECS: LW8600000
UN No: UN2803; Haz Class: 8; Packing Group: III
Low Melting Solid; Molecular Formula: Ga; MW: 69.72
Melting Point: 29.8°; Boiling Point: 2403°
Density (g/mL): 5.904
- RTECS: LW8600000
- Low Melting Solid
Formula:
Ga Boiling Pt: 2403 °C Melting Pt: 29.8 °C Density: 5.904 g/mL |
MDL Number:
MFCD00134045 CAS Number: 7440-55-3 EINECS: 231-163-8 UN: 2803 ADR: 8,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...