Chromium(II) chloride, anhydrous ≥97%

Supplier: BeanTown Chemical

Synonyms: Chromium dichloride

134290-100G 134290-25G 134290-5G
BT134290-100GEA 658.19 USD
BT134290-100G BT134290-25G BT134290-5G
Chromium(II) chloride, anhydrous ≥97%
Chromium(II) chloride

CAS: 10049-05-5; EC No: 233-163-3; MDL No: MFCD00010947; RTECS: GB5250000 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: CrCl2; MW: 122.90 Melting Point: 824° Density (g/mL): 2.9 Air Sensitive, Moisture Sensitive


  • RTECS: GB5250000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: Cl₂Cr
MW: 122.9 g/mol
Boiling Pt: 1302
Melting Pt: 824 °C
Density: 2.9 g/mL
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8 III
Merck Index: 13,02264

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR