Triethylsilyl trifluoromethanesulphonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Triethylsilyl trifluoromethanesulphonate

134280-25G 134280-5G
BT134280-25GEA 121 USD
BT134280-25G BT134280-5G
Triethylsilyl trifluoromethanesulphonate ≥98%
Triethylsilyl trifluoromethanesulphonate

CAS: 79271-56-0; EC No: 279-124-4; MDL No: MFCD00000407 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CF3SO3Si(CH2CH3)3; Molecular Formula: C7H15F3O3SSi; MW: 264.33 Boiling Point: 207°; Flash point: 66°C (151°F) Density (g/mL): 1.169; Refractive Index: 1.389 Moisture Sensitive


  • Refractive index: 1.389
  • Moisture Sensitive
  • Liquid

Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 207 °C
Density: 1.169 g/mL
Flash Pt: 66 °C (151 °F)
MDL Number: MFCD00000407
CAS Number: 79271-56-0
EINECS: 279-124-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR