Niobium(V) ethanolate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

134195-1G 134195-5G
BT134195-1GEA 44.26 USD
BT134195-1G BT134195-5G
Niobium(V) ethanolate ≥99.9% (trace metals basis)
Niobium(V) ethanolate

CAS: 3236-82-6; EC No: 221-795-2; MDL No: MFCD00015122 UN No: UN2924; Haz Class: 3(8); Packing Group: III Liquid; Linear Formula: Nb(OCH2CH3)5; MW: 318.21 Melting Point: 6°; Boiling Point: 142°/0.1 mmHg; Flash point: 36°C (97°F) Density (g/mL): 1.268; Refractive Index: 1.516 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.516
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₁₀H₂₅NbO₅
Boiling Pt: 142 °C/0.1 mmHg
Melting Pt: 6 °C
Density: 1.268 g/mL
Flash Pt: 36 °C (97 °F)
MDL Number: MFCD00015122
CAS Number: 3236-82-6
EINECS: 221-795-2
ADR: 3(8),III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR