Potassium permanganate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Permanganic acid potassium salt

134160-100G 134160-2KG 134160-500G
BT134160-100GEA 45.94 USD
BT134160-100G BT134160-2KG BT134160-500G
Potassium permanganate ≥99.0% ACS
Potassium permanganate

CAS: 7722-64-7; EC No: 231-760-3; MDL No: MFCD00011364; RTECS: SD6475000 UN No: UN1490; Haz Class: 5.1; Packing Group: II Crystalline; Molecular Formula: KMnO4; MW: 158.03 Melting Point: 240° Density (g/mL): 2.71 Light Sensitive


  • RTECS: SD6475000
  • Light Sensitive
  • Crystalline

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: 240 °C
Density: 2.71 g/mL
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR