1-Bromobutane ≥98%
Supplier: BeanTown Chemical
Synonyms:
Butyl bromide
134125-2.5KG
134125-500G
BT134125-10KGEA
352.59
USD
BT134125-10KG
BT134125-2.5KG
BT134125-500G
1-Bromobutane ≥98%
1-Bromobutane
CAS: 109-65-9; EC No: 203-691-9; MDL No: MFCD00000260; RTECS: EJ6225000
UN No: UN1126; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3(CH2)3Br; MW: 137.02
Melting Point: -112°; Boiling Point: 100-104°; Flash point: 10°C (50°F)
Density (g/mL): 1.276; Refractive Index: 1.439
- RTECS: EJ6225000
- Refractive index: 1.439
- Liquid
Formula:
BrH₂C(CH₂)₂CH₃ MW: 137.02 g/mol Boiling Pt: 100…104 °C Melting Pt: –112 °C Density: 1.276 g/mL Flash Pt: 10 °C (50 °F) |
MDL Number:
MFCD00000260 CAS Number: 109-65-9 EINECS: 203-691-9 UN: 1126 ADR: 3 II Merck Index: 13,01552 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...