2-Mercaptobenzimidazole ≥97%

Supplier: BeanTown Chemical

Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

134000-100G 134000-2.5KG 134000-500G
BT134000-100GEA 28.98 USD
BT134000-100G BT134000-2.5KG BT134000-500G
2-Mercaptobenzimidazole ≥97%
2-Mercaptobenzimidazole

CAS: 583-39-1; EC No: 209-502-6; MDL No: MFCD00466107; RTECS: DE1050000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C7H6N2S; MW: 150.20 Melting Point: 300-304°


  • RTECS: DE1050000
  • Crystalline

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 300…304 °C
MDL Number: MFCD00466107
CAS Number: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR