Cadmium chloride hydrate ≥99.995%
Supplier: BeanTown Chemical
133945-1G
133945-5G
BT133945-1GEA
71
USD
BT133945-1G
BT133945-5G
Cadmium chloride hydrate ≥99.995%
Cadmium chloride hydrate
CAS: 654054-66-7; EC No: 233-296-7; MDL No: MFCD00149627; RTECS: EV0175000
UN No: UN2570; Haz Class: 6.1; Packing Group: II
Crystalline/Powder; Molecular Formula: CdCl2·xH2O ; MW: 183.32 (anhydrous)
Hygroscopic
- RTECS: EV0175000
- Hygroscopic
- Crystalline/Powder
Formula:
CdCl₂·H₂O MW: 201.329 g/mol |
CAS Number:
654054-66-7 EINECS: 233-296-7 ADR: 6.1 II |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...