Methyl acetate ≥99%
Supplier: BeanTown Chemical
CAS: 79-20-9; EC No: 201-185-2; MDL No: MFCD00008711; RTECS: AI9100000
UN No: UN1231; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3COOCH3; MW: 74.08
Melting Point: -98°; Boiling Point: 57-58°; Flash point: -13°C (9°F)
Density (g/mL): 0.934; Refractive Index: 1.361
Moisture Sensitive
- RTECS: AI9100000
- Refractive index: 1.361
- Moisture Sensitive
- Liquid
Formula:
C₃H₆O₂ Boiling Pt: 57…58 °C Melting Pt: –98 °C Density: 0.934 g/mL Flash Pt: –13 °C (9 °F) |
MDL Number:
MFCD00008711 CAS Number: 79-20-9 EINECS: 201-185-2 UN: 1231 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...