N-Ethyldiisopropylamine ≥99%

Supplier: BeanTown Chemical

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA

133915-100ML 133915-2.5L 133915-500ML
BT133915-100MLEA 46.2 USD
BT133915-100ML BT133915-2.5L BT133915-500ML
N-Ethyldiisopropylamine ≥99%
N-Ethyldiisopropylamine

CAS: 7087-68-5; EC No: 230-392-0; MDL No: MFCD00008868 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: [(CH3)2CH]2NCH2CH3; Molecular Formula: C8H19N; MW: 129.25 Melting Point: -50°; Boiling Point: 126-127°; Flash point: 8°C (46°F) Density (g/mL): 0.742; Refractive Index: 1.414


  • Refractive index: 1.414
  • Liquid

Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127
Melting Pt: -50
Density: 0.742 g/mL
Flash Pt: 8 °C (46 °F)
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR