(1-Butyl)triphenylphosphonium bromide ≥99%

Supplier: BeanTown Chemical

Synonyms: Butyltriphenylphosphonium bromide

133795-100G 133795-25G 133795-500G
BT133795-100GEA 83 USD
BT133795-100G BT133795-25G BT133795-500G
(1-Butyl)triphenylphosphonium bromide ≥99%
(1-Butyl)triphenylphosphonium bromide

CAS: 1779-51-7; EC No: 217-219-4; MDL No: MFCD00011855; RTECS: TA1855200 UN No: UN3464; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: (C6H5)3P(Br)CH2CH2CH2CH3; Molecular Formula: C22H24BrP; MW: 339.30 Melting Point: 240-243° Hygroscopic


  • RTECS: TA1855200
  • Hygroscopic
  • Crystalline

Formula: H₃C(CH₂)₃P(C₆H₅)₃Br
MW: 399.31 g/mol
Melting Pt: 240…243 °C
MDL Number: MFCD00011855
CAS Number: 1779-51-7
EINECS: 217-219-4
UN: 3464
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e322e1de83c6056c1da2768b0235261af439bb0&vid=2e322e1de83c6056c1da2768b0235261af439bb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR