Sulphuric acid 1.0 N in water volumetric solution
Supplier: BeanTown Chemical
133790-1L
133790-20L
133790-4L
BT133790-1LEA
47.59
USD
BT133790-1L
BT133790-20L
BT133790-4L
Sulphuric acid 1.0 N in water volumetric solution
Sulfuric acid
CAS: 7664-93-9; EC No: 231-639-5; MDL No: MFCD00064589; RTECS: WS5600000
UN No: UN2796; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: H2SO4; MW: 98.08
Density (g/mL): 1.014
- RTECS: WS5600000
- Liquid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.014 g/mL |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...