2,2-Dimethyl-1-propanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Neopentyl alcohol

133785-100G 133785-25G 133785-500G
BT133785-100GEA 150 USD
BT133785-100G BT133785-25G BT133785-500G
2,2-Dimethyl-1-propanol ≥99%
2,2-Dimethyl-1-propanol

CAS: 75-84-3; EC No: 200-907-3; MDL No: MFCD00004682 UN No: UN1325; Haz Class: 4.1; Packing Group: II Solid; Linear Formula: (CH3)3CCH2OH; Molecular Formula: C5H12O; MW: 88.15 Melting Point: 52-56°; Boiling Point: 111-114°; Flash point: 36°C (96°F) Density (g/mL): 0.818


  • Solid

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 111…114 °C
Melting Pt: 52…56 °C
Density: 0.818 g/mL
Flash Pt: 36 °C (96 °F)
MDL Number: MFCD00004682
CAS Number: 75-84-3
EINECS: 200-907-3
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR