Mercury(II) iodide ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Mercury diiodide, Mercuric iodide

133740-100G 133740-500G
BT133740-100GEA 106.27 USD
BT133740-100G BT133740-500G
Mercury(II) iodide ≥99.0% ACS
Mercury(II) iodide

CAS: 7774-29-0; EC No: 231-873-8; MDL No: MFCD00011044; RTECS: OW5250000 UN No: UN1638; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: HgI2; MW: 454.40 Melting Point: 259°; Boiling Point: 354° Density (g/mL): 6.36 Light Sensitive


  • RTECS: OW5250000
  • Light Sensitive
  • Powder

Formula: HgI₂
MW: 454.4 g/mol
Boiling Pt: 354
Melting Pt: 259
Density: 6.36 g/mL
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1 II
Merck Index: 13,05905

Order Now

Specification Test Results

Assay 99.0% min.
Mercurous mercury (as Hg) ≤0.1%
Soluble mercury salts (as Hg) ≤0.05%
Solubility in potassium iodide Passes test
Description Red to orange-red powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR